Assay Details
| Gene Symbol: TMEM14C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087822.4 | XP_001087822.1 | 550 | 64 | ||
| XM_015135526.2 | XP_014991012.1 | 564 | 64 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM14C
Genomic Map