Assay Details
| Gene Symbol: CHMP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836900.1 | XP_028692733.1 | 292 | 125 | ||
| XM_028836901.1 | XP_028692734.1 | 247 | 125 | |||
| GenBank mRNA | JU336389.1 | - | 190 | 125 | ||
| JU477210.1 | - | 202 | 125 | |||
| JV048256.1 | - | 203 | 125 | |||
| JV566182.1 | - | 208 | 125 | |||
More Information
Set Membership: |
|

CHMP6
Genomic Map