Assay Details
| Gene Symbol: CLDN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114708.4 | XP_001114708.1 | 620 | 76 | ||
| XM_015132093.2 | XP_014987579.1 | 844 | 76 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CLDN18
Genomic Map