Assay Details
Gene Symbol: CWC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266625.1 | NP_001253554.1 | 346 | 93 | ||
XM_015115449.2 | XP_014970935.1 | 373 | 93 | |||
GenBank mRNA | JU328134.1 | - | 234 | 93 | ||
JU475916.1 | - | 252 | 93 | |||
JV043389.1 | - | 346 | 93 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
