Assay Details
| Gene Symbol: ST8SIA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083950.1 | NP_001077419.1 | 669 | 57 | ||
| XM_015121872.2 | XP_014977358.1 | 4280 | 57 | |||
| XM_028837689.1 | XP_028693522.1 | 1081 | 57 | |||
| GenBank mRNA | AM491802.1 | - | 669 | 57 | ||
| JV529282.1 | - | 705 | 57 | |||
| JV564989.1 | - | 965 | 57 | |||
| JV681916.1 | - | 1089 | 57 | |||
More Information
Set Membership: |
|

ST8SIA5
Genomic Map