Assay Details
| Gene Symbol: CHMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112797.2 | XP_014968283.1 | 320 | 128 | ||
| GenBank mRNA | JU334951.1 | - | 222 | 128 | ||
| JU475960.1 | - | 228 | 128 | |||
| JU475961.1 | - | 228 | 128 | |||
| JV045539.1 | - | 245 | 128 | |||
| JV716733.1 | - | 248 | 128 | |||
| JV716734.1 | - | 248 | 128 | |||
More Information
Set Membership: |
|

CHMP3
Genomic Map