Assay Details
| Gene Symbol: TMEM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266204.1 | NP_001253133.1 | 358 | 64 | ||
| XM_015139531.2 | XP_014995017.2 | 443 | 64 | |||
| GenBank mRNA | JU337211.1 | - | 380 | 64 | ||
| JU477926.1 | - | 248 | 64 | |||
| JV048769.1 | - | 358 | 64 | |||
| JV600737.1 | - | 400 | 64 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM59
Genomic Map