Assay Details
| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119152.2 | XP_014974638.2 | 2944 | 65 | ||
| GenBank mRNA | JV637835.1 | - | 1207 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF43| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119152.2 | XP_014974638.2 | 2944 | 65 | ||
| GenBank mRNA | JV637835.1 | - | 1207 | 65 | ||
Set Membership: |
|