Assay Details
| Gene Symbol: GMFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143718.2 | XP_014999204.1 | 289 | 77 | ||
| XM_015143719.2 | XP_014999205.1 | 411 | 77 | |||
| XM_028851559.1 | XP_028707392.1 | 289 | 77 | |||
| GenBank mRNA | JU334056.1 | - | 171 | 77 | ||
| JU470505.1 | - | 240 | 77 | |||
| JV668471.1 | - | 251 | 77 | |||
| JV668472.1 | - | 251 | 77 | |||
More Information
Set Membership: |
|

GMFB
Genomic Map