Assay Details
| Gene Symbol: TMEM106B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193700.2 | NP_001180629.1 | 438 | 72 | ||
| XM_002803370.3 | XP_002803416.1 | 368 | 72 | |||
| XM_015133985.2 | XP_014989471.1 | 469 | 72 | |||
| XM_015133986.2 | XP_014989472.1 | 433 | 72 | |||
| GenBank mRNA | JU321942.1 | - | 334 | 72 | ||
| JU471780.1 | - | 269 | 72 | |||
| JV044767.1 | - | 216 | 72 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM106B
Genomic Map