Assay Details
| Gene Symbol: NPY1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032866.1 | NP_001028038.1 | 690 | 127 | ||
| XM_015139391.2 | XP_014994877.1 | 1839 | 127 | |||
| XM_015139392.2 | XP_014994878.1 | 931 | 127 | |||
| GenBank mRNA | AF303089.1 | - | 690 | 127 | ||
| JU334697.1 | - | 928 | 127 | |||
| JV469862.1 | - | 940 | 127 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NPY1R
Genomic Map