Assay Details
| Gene Symbol: ATXN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260745.1 | NP_001247674.1 | 865 | 87 | ||
| GenBank mRNA | JU333849.1 | - | 942 | 87 | ||
| JU333850.1 | - | 942 | 87 | |||
| JU472415.1 | - | 954 | 87 | |||
| JV046672.1 | - | 865 | 87 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ATXN10
Genomic Map