Assay Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112753.2 | XP_014968239.1 | 1108 | 76 | ||
| XM_015112754.2 | XP_014968240.1 | 1115 | 76 | |||
More Information
Set Membership: |
|

TRABD2A
Genomic Map