Assay Details
| Gene Symbol: ASF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194486.1 | NP_001181415.1 | 448 | 78 | ||
| GenBank mRNA | JU319812.1 | - | 359 | 78 | ||
| JU471032.1 | - | 401 | 78 | |||
| JV043846.1 | - | 369 | 78 | |||
| JV638957.1 | - | 455 | 78 | |||
More Information
Set Membership: |
|

ASF1A
Genomic Map