Assay Details
| Gene Symbol: MPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203259.1 | NP_001190188.1 | 336 | 62 | ||
| XM_028840597.1 | XP_028696430.1 | 1037 | 62 | |||
| GenBank mRNA | CN643206.1 | - | 30 | 62 | ||
| CO583027.1 | - | 340 | 62 | |||
| JU343798.1 | - | 323 | 62 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MPG
Genomic Map