Assay Details
| Gene Symbol: ZSCAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135772.2 | XP_014991258.1 | 655 | 89 | ||
| GenBank mRNA | JU328483.1 | - | 268 | 89 | ||
| JV521425.1 | - | 520 | 89 | |||
More Information
Set Membership: |
|

ZSCAN16
Genomic Map