Assay Details
| Gene Symbol: ZNF613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124717.2 | XP_014980203.2 | 589 | 94 | ||
| XM_015124718.2 | XP_014980204.2 | 2178 | 94 | |||
| XM_015124719.2 | XP_014980205.2 | 586 | 94 | |||
| XM_015124721.2 | XP_014980207.2 | 507 | 94 | |||
| XM_015124722.2 | XP_014980208.2 | 171 | 94 | |||
| XM_028839359.1 | XP_028695192.1 | 297 | 94 | |||
| GenBank mRNA | JU493718.1 | - | 213 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015124717.2, XM_015124718.2, XM_015124719.2, XM_015124721.2, XM_015124722.2, XM_028839359.1 transcripts.
Set Membership: |
|

ZNF613
Genomic Map