Assay Details
| Gene Symbol: TRMT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261660.1 | NP_001248589.1 | 367 | 129 | ||
| GenBank mRNA | JU321196.1 | - | 363 | 129 | ||
| JU475941.1 | - | 367 | 129 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TRMT6
Genomic Map