Assay Details
| Gene Symbol: SNAPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257650.1 | NP_001244579.1 | 262 | 72 | ||
| GenBank mRNA | JU333731.1 | - | 262 | 72 | ||
| JV046610.1 | - | 220 | 72 | |||
| JV462926.1 | - | 147 | 72 | |||
| JV647038.1 | - | 262 | 72 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SNAPIN
Genomic Map