Assay Details
| Gene Symbol: EFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142866.2 | XP_014998352.1 | 337 | 64 | ||
| XM_028851229.1 | XP_028707062.1 | 337 | 64 | |||
| GenBank mRNA | JU477209.1 | - | 270 | 64 | ||
| JV637621.1 | - | 267 | 64 | |||
More Information
Set Membership: |
|

EFL1
Genomic Map