Assay Details
| Gene Symbol: BARX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835358.1 | XP_028691191.1 | 812 | 70 | ||
| GenBank mRNA | JV065869.1 | - | 215 | 70 | ||
| JV656116.1 | - | 318 | 70 | |||
More Information
Set Membership: |
|

BARX1
Genomic Map