Assay Details
| Gene Symbol: MDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114947.1 | NP_001108419.1 | 139 | 76 | ||
| GenBank mRNA | JU496856.1 | - | 239 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MDC1| Gene Symbol: MDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114947.1 | NP_001108419.1 | 139 | 76 | ||
| GenBank mRNA | JU496856.1 | - | 239 | 76 | ||
Set Membership: |
|