Assay Details
| Gene Symbol: SLC22A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271646.1 | NP_001258575.1 | 1284 | 115 | ||
| XM_015113722.2 | XP_014969208.2 | 1737 | 115 | |||
| XM_015113724.2 | XP_014969210.2 | 1635 | 115 | |||
| XM_015113725.2 | XP_014969211.2 | 1422 | 115 | |||
| XM_028832811.1 | XP_028688644.1 | 1267 | 115 | |||
| XM_028832812.1 | XP_028688645.1 | 1027 | 115 | |||
| GenBank mRNA | AB738913.1 | - | 1284 | 115 | ||
More Information
Set Membership: |
|

SLC22A12
Genomic Map