Assay Details
| Gene Symbol: YIPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260624.1 | NP_001247553.1 | 636 | 78 | ||
| XM_015136369.2 | XP_014991855.1 | 715 | 78 | |||
| XM_015136370.2 | XP_014991856.1 | 710 | 78 | |||
| GenBank mRNA | JU321782.1 | - | 578 | 78 | ||
| JU470971.1 | - | 586 | 78 | |||
| JV043270.1 | - | 636 | 78 | |||
| JV566201.1 | - | 615 | 78 | |||
More Information
Set Membership: |
|

YIPF3
Genomic Map