Assay Details
| Gene Symbol: RTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143801.2 | XP_014999287.2 | 2617 | 89 | ||
| XM_015143802.2 | XP_014999288.1 | 820 | 89 | |||
| XM_028851579.1 | XP_028707412.1 | 534 | 89 | |||
| GenBank mRNA | JV565478.1 | - | 2397 | 89 | ||
| JV590136.1 | - | 632 | 89 | |||
| JV667172.1 | - | 2338 | 89 | |||
| JV667451.1 | - | 766 | 89 | |||
More Information
Set Membership: |
|

RTN1
Genomic Map