Assay Details
| Gene Symbol: TMEM150A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087786.4 | XP_001087786.2 | 616 | 53 | ||
| XM_001088263.4 | XP_001088263.2 | 603 | 53 | |||
| XM_015112765.2 | XP_014968251.2 | 823 | 53 | |||
| XM_015112768.2 | XP_014968254.2 | 549 | 53 | |||
| XM_015112769.2 | XP_014968255.2 | 536 | 53 | |||
| XM_028832331.1 | XP_028688164.1 | 760 | 53 | |||
| GenBank mRNA | JU473038.1 | - | 519 | 53 | ||
| JV635841.1 | - | 575 | 53 | |||
More Information
Set Membership: |
|

TMEM150A
Genomic Map