Assay Details
| Gene Symbol: TMEM95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194382.2 | NP_001181311.2 | 571 | 73 | ||
| XM_015118454.2 | XP_014973940.2 | 597 | 73 | |||
| XM_028835691.1 | XP_028691524.1 | 614 | 73 | |||
| GenBank mRNA | JU342755.1 | - | 316 | 73 | ||
More Information
Set Membership: |
|

TMEM95
Genomic Map