Assay Details
| Gene Symbol: AP2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119081.2 | XP_014974567.1 | 825 | 89 | ||
| XM_015119083.2 | XP_014974569.1 | 622 | 89 | |||
| XM_015119084.2 | XP_014974570.1 | 348 | 89 | |||
| XM_015119085.2 | XP_014974571.1 | 355 | 89 | |||
| XM_015119088.2 | XP_014974574.1 | 467 | 89 | |||
| XM_028836207.1 | XP_028692040.1 | 453 | 89 | |||
| GenBank mRNA | JU333185.1 | - | 420 | 89 | ||
| JU333186.1 | - | 420 | 89 | |||
| JU333187.1 | - | 420 | 89 | |||
| JU474500.1 | - | 429 | 89 | |||
| JU474501.1 | - | 429 | 89 | |||
| JV046311.1 | - | 467 | 89 | |||
| JV046312.1 | - | 467 | 89 | |||
| JV708268.1 | - | 472 | 89 | |||
More Information
Set Membership: |
|

AP2B1
Genomic Map