Assay Details
| Gene Symbol: LCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266344.1 | NP_001253273.1 | 1144 | 88 | ||
| XM_015136779.2 | XP_014992265.2 | 1342 | 88 | |||
| XM_015136780.2 | XP_014992266.2 | 1419 | 88 | |||
| XM_015136783.2 | XP_014992269.2 | 1470 | 88 | |||
| XM_028846849.1 | XP_028702682.1 | 1547 | 88 | |||
| XM_028846850.1 | XP_028702683.1 | 1290 | 88 | |||
| GenBank mRNA | JU333211.1 | - | 1144 | 88 | ||
| JV703816.1 | - | 1388 | 88 | |||
More Information
Set Membership: |
|

LCA5
Genomic Map