Assay Details
| Gene Symbol: BTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101880.4 | XP_001101880.3 | 647 | 79 | ||
| GenBank mRNA | JV048060.1 | - | 583 | 79 | ||
| JV636341.1 | - | 605 | 79 | |||
More Information
Set Membership: |
|

BTC
Genomic Map