Assay Details
| Gene Symbol: SHCBP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122209.2 | XP_014977695.1 | 1563 | 92 | ||
| XM_015122216.2 | XP_014977702.1 | 1563 | 92 | |||
More Information
Set Membership: |
|

SHCBP1L
Genomic Map