Assay Details
Gene Symbol: FAM107B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146517.2 | XP_015002003.1 | 344 | 86 | ||
XM_015146519.2 | XP_015002005.1 | 448 | 86 | |||
XM_028826005.1 | XP_028681838.1 | 361 | 86 | |||
GenBank mRNA | JU349236.1 | - | 191 | 86 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
