Assay Details
| Gene Symbol: NGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148902.2 | XP_015004388.1 | 193 | 78 | ||
| GenBank mRNA | JV062030.1 | - | 136 | 78 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_015148902.2 transcripts.
This assay is in the 5'UTR region of the XM_015148902.2 transcripts.
Set Membership: |
|

NGF
Genomic Map