Assay Details
| Gene Symbol: RILPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260833.1 | NP_001247762.1 | 582 | 57 | ||
| XM_028828873.1 | XP_028684706.1 | 866 | 57 | |||
| XM_028828874.1 | XP_028684707.1 | 866 | 57 | |||
| GenBank mRNA | JU333278.1 | - | 652 | 57 | ||
| JU474589.1 | - | 603 | 57 | |||
| JV046372.1 | - | 582 | 57 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RILPL2
Genomic Map