Assay Details
| Gene Symbol: EPHX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258087.1 | NP_001245016.1 | 258 | 89 | ||
| XM_015114872.2 | XP_014970358.1 | 434 | 89 | |||
| XM_028834554.1 | XP_028690387.1 | 875 | 89 | |||
| GenBank mRNA | JU333345.1 | - | 250 | 89 | ||
| JU474645.1 | - | 258 | 89 | |||
| JV046427.1 | - | 258 | 89 | |||
| JV565077.1 | - | 247 | 89 | |||
| JV709216.1 | - | 253 | 89 | |||
More Information
Set Membership: |
|

EPHX1
Genomic Map