Assay Details
| Gene Symbol: AHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808282.3 | XP_002808328.3 | 6118 | 61 | ||
| XM_015132780.2 | XP_014988266.2 | 6013 | 61 | |||
| XM_015132787.2 | XP_014988273.2 | 6132 | 61 | |||
| XM_015132795.2 | XP_014988281.2 | 6010 | 61 | |||
| XM_015132800.2 | XP_014988286.2 | 5788 | 61 | |||
| XM_015132808.2 | XP_014988294.2 | 5893 | 61 | |||
| XM_015132818.2 | XP_014988304.2 | 6050 | 61 | |||
| XM_028837300.1 | XP_028693133.1 | 5606 | 61 | |||
| XM_028837308.1 | XP_028693141.1 | 6018 | 61 | |||
| XM_028837315.1 | XP_028693148.1 | 5785 | 61 | |||
| GenBank mRNA | JV541322.1 | - | 1677 | 61 | ||
| JV600268.1 | - | 5093 | 61 | |||
More Information
Set Membership: |
|

AHDC1
Genomic Map