Assay Details
| Gene Symbol: RIMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261567.1 | NP_001248496.1 | 857 | 95 | ||
| XM_015136163.2 | XP_014991649.1 | 922 | 95 | |||
| XM_028827492.1 | XP_028683325.1 | 1121 | 95 | |||
| XM_028827578.1 | XP_028683411.1 | 1102 | 95 | |||
| GenBank mRNA | JU319693.1 | - | 857 | 95 | ||
| JV667978.1 | - | 1007 | 95 | |||
More Information
Set Membership: |
|

RIMS3
Genomic Map