Assay Details
| Gene Symbol: NOSIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194679.2 | NP_001181608.2 | 287 | 56 | ||
| XM_015124396.2 | XP_014979882.2 | 301 | 56 | |||
| GenBank mRNA | JU322037.1 | - | 183 | 56 | ||
| JU471600.1 | - | 187 | 56 | |||
| JV452578.1 | - | 178 | 56 | |||
More Information
Set Membership: |
|

NOSIP
Genomic Map