Assay Details
Gene Symbol: GSTCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266390.1 | NP_001253319.1 | 1588 | 67 | ||
XM_015138905.2 | XP_014994391.1 | 1802 | 67 | |||
GenBank mRNA | JU322464.1 | - | 1588 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
