Assay Details
| Gene Symbol: PDZRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151453.2 | XP_015006939.1 | 1693 | 102 | ||
| XM_015151454.2 | XP_015006940.1 | 605 | 102 | |||
| GenBank mRNA | JU342516.1 | - | 655 | 102 | ||
More Information
Set Membership: |
|

PDZRN4
Genomic Map