Assay Details
| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194079.2 | NP_001181008.1 | 856 | 81 | ||
| XM_015133858.2 | XP_014989344.1 | 928 | 81 | |||
| XM_015133859.2 | XP_014989345.1 | 757 | 81 | |||
| GenBank mRNA | JU325733.1 | - | 785 | 81 | ||
| JV635836.1 | - | 879 | 81 | |||
More Information
Set Membership: |
|

FAM221A
Genomic Map