Assay Details
| Gene Symbol: CARMIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143420.2 | XP_014998906.1 | 1670 | 81 | ||
| XM_015143421.2 | XP_014998907.1 | 1670 | 81 | |||
| XM_015143422.2 | XP_014998908.1 | 1670 | 81 | |||
| XM_028851435.1 | XP_028707268.1 | 1670 | 81 | |||
| XM_028851436.1 | XP_028707269.1 | 1670 | 81 | |||
| XM_028851437.1 | XP_028707270.1 | 1670 | 81 | |||
| XM_028851438.1 | XP_028707271.1 | 1438 | 81 | |||
| GenBank mRNA | JV726512.1 | - | 1709 | 81 | ||
More Information
Set Membership: |
|

CARMIL3
Genomic Map