Assay Details
| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261601.1 | NP_001248530.1 | 2881 | 113 | ||
| XM_028830360.1 | XP_028686193.1 | 3074 | 113 | |||
| XM_028830361.1 | XP_028686194.1 | 2989 | 113 | |||
| XM_028830362.1 | XP_028686195.1 | 3124 | 113 | |||
| XM_028830363.1 | XP_028686196.1 | 2923 | 113 | |||
| XM_028830364.1 | XP_028686197.1 | 3372 | 113 | |||
| GenBank mRNA | JU321255.1 | - | 2973 | 113 | ||
| JV043405.1 | - | 2881 | 113 | |||
| JV638180.1 | - | 2975 | 113 | |||
More Information
Set Membership: |
|

SH3BP4
Genomic Map