Assay Details
| Gene Symbol: PLGRKT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257744.1 | NP_001244673.1 | 517 | 83 | ||
| XM_028834207.1 | XP_028690040.1 | 616 | 83 | |||
| GenBank mRNA | JU322534.1 | - | 517 | 83 | ||
| JU473365.1 | - | 536 | 83 | |||
| JV045409.1 | - | 419 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PLGRKT
Genomic Map