Assay Details
Gene Symbol: ZNF827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139301.2 | XP_014994787.1 | 2040 | 67 | ||
XM_015139303.2 | XP_014994789.1 | 2040 | 67 | |||
XM_015139304.2 | XP_014994790.1 | 2040 | 67 | |||
XM_015139305.2 | XP_014994791.1 | 1681 | 67 | |||
XM_015139306.2 | XP_014994792.1 | 2040 | 67 | |||
XM_028848786.1 | XP_028704619.1 | 2040 | 67 | |||
XM_028848787.1 | XP_028704620.1 | 1809 | 67 | |||
XM_028848788.1 | XP_028704621.1 | 2040 | 67 | |||
XM_028848789.1 | XP_028704622.1 | 2040 | 67 | |||
GenBank mRNA | JU331220.1 | - | 320 | 67 | ||
JV643424.1 | - | 205 | 67 | |||
JV707908.1 | - | 1285 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
