Assay Details
| Gene Symbol: EGR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144966.2 | XP_015000452.1 | 674 | 70 | ||
| GenBank mRNA | HM370983.1 | - | 393 | 70 | ||
| JU333276.1 | - | 164 | 70 | |||
More Information
Set Membership: |
|

EGR3
Genomic Map