Assay Details
| Gene Symbol: XKRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090721.4 | XP_001090721.1 | 1849 | 73 | ||
| GenBank mRNA | JU486597.1 | - | 384 | 73 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

XKRX
Genomic Map