Assay Details
| Gene Symbol: LHFPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194055.1 | NP_001180984.1 | 684 | 73 | ||
| XM_015130314.2 | XP_014985800.1 | 628 | 73 | |||
| GenBank mRNA | JU328040.1 | - | 329 | 73 | ||
| JV529784.1 | - | 421 | 73 | |||
| JV581071.1 | - | 449 | 73 | |||
| JV683240.1 | - | 446 | 73 | |||
More Information
Set Membership: |
|

LHFPL4
Genomic Map