Assay Details
| Gene Symbol: TLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130425.1 | NP_001123897.1 | 1964 | 114 | ||
| XM_015139351.2 | XP_014994837.2 | 2177 | 114 | |||
| XM_015139352.2 | XP_014994838.2 | 2061 | 114 | |||
| GenBank mRNA | AB445630.1 | - | 1964 | 114 | ||
| AY525612.1 | - | 47 | 114 | |||
| EU204933.1 | - | 2023 | 114 | |||
More Information
Set Membership: |
|

TLR2
Genomic Map