Assay Details
Gene Symbol: TICAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY530100.1 | - | 357 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
